Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF6
Entrez Gene ID
487151
Gene Name
Kruppel like factor 6
Gene Aliases
-
Chromosome Location
Chr.2: 31654142 - 31664009 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544279.6 XP_544279.3 - 936 142

View Full Product Details