Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HES6
Entrez Gene ID
607298
Gene Name
hes family bHLH transcription factor 6
Gene Aliases
-
Chromosome Location
Chr.25: 48710694 - 48713392 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_843923.3 XP_849016.2 2 - 3 397 149

View Full Product Details