Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1QTNF6
Entrez Gene ID
481273
Gene Name
C1q and TNF related 6
Gene Aliases
-
Chromosome Location
Chr.10: 27340547 - 27349183 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538394.6 XP_538394.3 2 - 3 544 58

View Full Product Details