Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KLK6
Entrez Gene ID
476402
Gene Name
kallikrein related peptidase 6
Gene Aliases
-
Chromosome Location
Chr.1: 105902147 - 105907585 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197166.1 NP_001184095.1 - 451 169
GenBank mRNA HM135454.1 - - 451 169

View Full Product Details