Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCLAF3
Entrez Gene ID
611699
Gene Name
BCLAF1 and THRAP3 family member 3
Gene Aliases
CXHXorf23
Chromosome Location
Chr.X: 15844064 - 15903535 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014111742.2 XP_013967217.1 6 - 6 1618 98
XM_014111743.2 XP_013967218.1 6 - 6 1639 98
XM_014111744.2 XP_013967219.1 6 - 6 1618 98
XM_022415671.1 XP_022271379.1 4 - 4 1528 98
XM_022415672.1 XP_022271380.1 6 - 6 1618 98

View Full Product Details