Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AEBP2
Entrez Gene ID
486656
Gene Name
AE binding protein 2
Gene Aliases
-
Chromosome Location
Chr.27: 27654141 - 27722922 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108309.1 XP_013963784.1 4 - 5 1001 149

View Full Product Details