Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDH20
Entrez Gene ID
100686939
Gene Name
protocadherin 20
Gene Aliases
-
Chromosome Location
Chr.22: 17150498 - 17156091 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005633953.3 XP_005634010.1 2 - 2 2417 182

View Full Product Details