Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCHR1
Entrez Gene ID
403563
Gene Name
melanin concentrating hormone receptor 1
Gene Aliases
GPR24
Chromosome Location
Chr.10: 24526687 - 24560948 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003038.1 NP_001003038.1 2 - 2 899 116
XM_005625672.3 XP_005625729.1 3 - 3 1201 116
XM_005625673.3 XP_005625730.1 2 - 2 1760 116
XM_022423977.1 XP_022279685.1 4 - 4 1345 116
XM_022423978.1 XP_022279686.1 2 - 2 1083 116
GenBank mRNA AY112658.1 - 2 - 2 899 116

View Full Product Details