Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC28A
Entrez Gene ID
476220
Gene Name
coiled-coil domain containing 28A
Gene Aliases
-
Chromosome Location
Chr.1: 31044102 - 31058173 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014112356.2 XP_013967831.1 - 740 78

View Full Product Details