Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
PPCS
Entrez Gene ID
475309
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
-
Chromosome Location
Chr.15: 813435 - 817732 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022427724.1 XP_022283432.1 3 - 3 213 135
XM_532539.4 XP_532539.1 3 - 3 668 135

View Full Product Details