Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINGO1
Entrez Gene ID
487660
Gene Name
leucine rich repeat and Ig domain containing 1
Gene Aliases
-
Chromosome Location
Chr.30: 38668199 - 38738178 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005638642.3 XP_005638699.1 4 - 4 1775 84
XM_022412599.1 XP_022268307.1 3 - 3 1574 84
XM_544785.6 XP_544785.2 2 - 2 1590 84

View Full Product Details