Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDX1
Entrez Gene ID
493994
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
IPF1, PDX-1
Chromosome Location
Chr.25: 11741863 - 11746885 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284471.2 NP_001271400.1 - 367 125
GenBank mRNA HQ454506.1 - - 367 125

View Full Product Details