Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP27C1
Entrez Gene ID
483869
Gene Name
cytochrome P450 family 27 subfamily C member 1
Gene Aliases
-
Chromosome Location
Chr.19: 23374295 - 23400054 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631884.3 XP_005631941.1 7 - 8 1062 112
XM_005631885.3 XP_005631942.1 7 - 8 1052 112
XM_005631886.3 XP_005631943.1 7 - 8 1075 112
XM_014121491.2 XP_013976966.1 7 - 8 1131 112

View Full Product Details