Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR13G1
Entrez Gene ID
482211
Gene Name
olfactory receptor family 13 subfamily G member 1
Gene Aliases
cOR2AK3
Chromosome Location
Chr.14: 1233712 - 1234635 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_539330.3 XP_539330.3 1 - 1 639 185

View Full Product Details