Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO3
Entrez Gene ID
486662
Gene Name
LIM domain only 3
Gene Aliases
-
Chromosome Location
Chr.27: 30150311 - 30208286 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637101.3 XP_005637158.1 1 - 2 193 133

View Full Product Details