Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPATCH8
Entrez Gene ID
480500
Gene Name
G-patch domain containing 8
Gene Aliases
-
Chromosome Location
Chr.9: 18927894 - 19044989 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537619.6 XP_537619.2 - 585 112

View Full Product Details