Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGFRL1
Entrez Gene ID
102151155
Gene Name
fibroblast growth factor receptor like 1
Gene Aliases
-
Chromosome Location
Chr.3: 91499088 - 91508982 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618675.3 XP_005618732.2 8 - 9 2169 130

View Full Product Details