Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBA4
Entrez Gene ID
486334
Gene Name
crystallin beta A4
Gene Aliases
-
Chromosome Location
Chr.26: 20272962 - 20280368 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080897.1 NP_001074366.1 4 - 5 339 163
XM_005636022.3 XP_005636079.1 5 - 6 982 163
XM_005636023.3 XP_005636080.1 4 - 5 865 163
GenBank mRNA EF195231.1 - 4 - 5 339 163

View Full Product Details