Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCEAL1
Entrez Gene ID
481000
Gene Name
transcription elongation factor A like 1
Gene Aliases
-
Chromosome Location
Chr.X: 77054291 - 77056337 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003435629.4 XP_003435677.1 2 - 2 506 134
XM_003435630.4 XP_003435678.1 2 - 2 486 134
XM_005641609.3 XP_005641666.1 2 - 2 486 134
XM_005641610.2 XP_005641667.1 3 - 3 572 134
XM_005641611.2 XP_005641668.1 3 - 3 568 134
XM_005641612.2 XP_005641669.1 3 - 3 547 134
XM_538121.5 XP_538121.1 3 - 3 581 134

View Full Product Details