Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMX2
Entrez Gene ID
610697
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
Chromosome Location
Chr.28: 28002966 - 28009955 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_848240.5 XP_853333.1 3 - 3 1445 77

View Full Product Details