Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNK6
Entrez Gene ID
484531
Gene Name
potassium two pore domain channel subfamily K member 6
Gene Aliases
-
Chromosome Location
Chr.1: 114665958 - 114677845 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616626.3 XP_005616683.1 1 - 1 456 111
XM_541645.6 XP_541645.2 1 - 1 462 111

View Full Product Details