Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN16
Entrez Gene ID
608218
Gene Name
claudin 16
Gene Aliases
-
Chromosome Location
Chr.34: 22392108 - 22414250 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845175.5 XP_850268.2 1 - 2 478 100

View Full Product Details