Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRAF2
Entrez Gene ID
480912
Gene Name
PRA1 domain family member 2
Gene Aliases
-
Chromosome Location
Chr.X: 42218038 - 42221075 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538032.6 XP_538032.1 1 - 1 337 54

View Full Product Details