Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSG2
Entrez Gene ID
612882
Gene Name
neuronal vesicle trafficking associated 2
Gene Aliases
HMP19
Chromosome Location
Chr.4: 38491995 - 38550672 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847624.5 XP_852717.1 2 - 3 335 124

View Full Product Details