Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR7
Entrez Gene ID
491743
Gene Name
toll like receptor 7
Gene Aliases
-
Chromosome Location
Chr.X: 9331243 - 9360461 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048124.2 NP_001041589.1 2 - 2 2931 124
XM_005641003.3 XP_005641060.1 4 - 4 3241 124
GenBank mRNA AB248956.1 - 2 - 2 2932 124
DQ333224.1 - 3 - 3 2970 124

View Full Product Details