Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
ALKBH5
Entrez Gene ID
609383
Gene Name
alkB homolog 5, RNA demethylase
Gene Aliases
-
Chromosome Location
Chr.5: 41339292 - 41365641 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846626.5 XP_851719.1 2 - 2 1876 122

View Full Product Details