Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRN3
Entrez Gene ID
482406
Gene Name
leucine rich repeat neuronal 3
Gene Aliases
-
Chromosome Location
Chr.14: 50893254 - 50928475 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628735.3 XP_005628792.1 3 - 3 2407 138
XM_539523.6 XP_539523.1 2 - 2 2253 138

View Full Product Details