Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX21
Entrez Gene ID
485900
Gene Name
sorting nexin family member 21
Gene Aliases
-
Chromosome Location
Chr.24: 33143459 - 33149418 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635122.3 XP_005635179.1 3 - 4 492 128

View Full Product Details