Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKIL
Entrez Gene ID
488159
Gene Name
SKI like proto-oncogene
Gene Aliases
-
Chromosome Location
Chr.34: 34871770 - 34898681 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005639910.3 XP_005639967.1 2 - 2 1318 134
XM_005639911.3 XP_005639968.1 2 - 2 1024 134

View Full Product Details