Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHX4
Entrez Gene ID
490300
Gene Name
LIM homeobox 4
Gene Aliases
-
Chromosome Location
Chr.7: 13779574 - 13825650 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547420.6 XP_547420.2 - 646 97

View Full Product Details