Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DACT3
Entrez Gene ID
612178
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
-
Chromosome Location
Chr.1: 109247743 - 109264655 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116573.2 XP_013972048.2 3 - 4 1473 142

View Full Product Details