Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TP53INP2
Entrez Gene ID
610064
Gene Name
tumor protein p53 inducible nuclear protein 2
Gene Aliases
-
Chromosome Location
Chr.24: 23768208 - 23801268 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005634952.3 XP_005635009.1 3 - 3 1326 113
XM_005634953.3 XP_005635010.1 3 - 3 484 113
XM_005634955.3 XP_005635012.1 3 - 3 724 113
XM_022409384.1 XP_022265092.1 4 - 4 1436 113
XM_022409385.1 XP_022265093.1 3 - 3 732 113
XM_022409386.1 XP_022265094.1 3 - 3 662 113

View Full Product Details