Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX6
Entrez Gene ID
488328
Gene Name
glutathione peroxidase 6
Gene Aliases
-
Chromosome Location
Chr.35: 25620843 - 25630288 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256320.1 NP_001243249.1 5 - 5 515 134

View Full Product Details