Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPTX2
Entrez Gene ID
100855792
Gene Name
neuronal pentraxin 2
Gene Aliases
-
Chromosome Location
Chr.6: 10805564 - 10816587 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639083.4 XP_003639131.1 - 393 149

View Full Product Details