Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUT7
Entrez Gene ID
449026
Gene Name
fucosyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.9: 48604879 - 48609292 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005379.1 NP_001005379.1 1 - 1 971 89
XM_005625017.3 XP_005625074.1 3 - 3 1539 89
XM_005625018.3 XP_005625075.1 3 - 3 2172 89
XM_022422564.1 XP_022278272.1 3 - 3 1281 89
GenBank mRNA AJ831837.1 - 1 - 1 971 89

View Full Product Details