Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR7A74
Entrez Gene ID
484883
Gene Name
olfactory receptor family 7 subfamily A member 74
Gene Aliases
CFOLF4
Chromosome Location
Chr.20: 47499997 - 47500926 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013422.1 NP_001013440.1 1 - 1 419 72

View Full Product Details