Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT10
Entrez Gene ID
491006
Gene Name
keratin 10
Gene Aliases
CK-10, K10, Ker10
Chromosome Location
Chr.9: 21863715 - 21868587 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116588.2 XP_013972063.1 - 650 96
XM_014116589.2 XP_013972064.1 - 650 96
XM_014116590.2 XP_013972065.1 - 650 96

View Full Product Details