Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRCD
Entrez Gene ID
100049006
Gene Name
photoreceptor disc component
Gene Aliases
-
Chromosome Location
Chr.9: 4179757 - 4190658 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097560.2 NP_001091029.1 - 184 136
XM_022422503.1 XP_022278211.1 - 2065 136
GenBank mRNA DQ390330.1 - - 184 136

View Full Product Details