Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R40
Entrez Gene ID
608842
Gene Name
taste 2 receptor member 40
Gene Aliases
-
Chromosome Location
Chr.16: 6439097 - 6440017 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145495.1 NP_001138967.1 - 440 79

View Full Product Details