Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAFA-T2R2
Entrez Gene ID
100271741
Gene Name
bitter taste receptor Cafa-T2R2
Gene Aliases
-
Chromosome Location
Chr.14: 27345807 - 27346718 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145504.1 NP_001138976.1 1 - 1 855 74

View Full Product Details