Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM6
Entrez Gene ID
100568301
Gene Name
tripartite motif containing 6
Gene Aliases
-
Chromosome Location
Chr.21: 28595887 - 28611562 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205190.2 NP_001192119.1 3 - 4 750 93
XM_005633260.3 XP_005633317.1 4 - 5 901 93
XM_014122406.2 XP_013977881.1 4 - 5 896 90
XM_022407407.1 XP_022263115.1 5 - 6 1058 93

View Full Product Details