Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFDN5
Entrez Gene ID
607260
Gene Name
prefoldin subunit 5
Gene Aliases
-
Chromosome Location
Chr.27: 1906133 - 1909613 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251949.1 NP_001238878.1 2 - 3 203 75
GenBank mRNA DN269529.1 - 2 - 3 188 75

View Full Product Details