Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MGST3
Entrez Gene ID
609050
Gene Name
microsomal glutathione S-transferase 3
Gene Aliases
-
Chromosome Location
Chr.38: 17670329 - 17691822 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252410.1 NP_001239339.1 3 - 4 292 105
GenBank mRNA DN433890.1 - 3 - 4 269 105
DT536092.1 - 3 - 4 359 105

View Full Product Details