Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR10B
- Entrez Gene ID
-
100886149
- Gene Name
- microRNA mir-10b
- Gene Aliases
-
cfa-mir-10b
- Chromosome Location
-
Chr.36: 19958913 - 19959013 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_049424.1 |
|
1 - 1 |
51 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
homeobox D8 |
HOXD8 |
|
-17.7kb
|
Sense |
Chr. - 19938916 - 19941092 on Build CanFam3.1 |
Downstream |
homeobox D3 |
HOXD3 |
|
2.5kb
|
Sense |
Chr. - 19961393 - 19981672 on Build CanFam3.1 |
View Full Product Details