Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ap3m1
Entrez Gene ID
100689277
Gene Name
adaptor-related protein complex 3, mu 1 subunit
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613632: 449388 - 470992 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244405.1 NP_001231334.1 6 - 7 1009 78
XM_007652612.1 XP_007650802.1 7 - 8 1046 78
XM_007652618.1 XP_007650808.1 7 - 8 1176 78
XM_007652623.1 XP_007650813.1 6 - 7 910 78
GenBank mRNA AY346109.1 - 7 - 8 1276 78
JI885336.1 - 7 - 8 1367 78
JP040800.1 - 7 - 8 1049 78

View Full Product Details