Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Selm
Entrez Gene ID
100762068
Gene Name
selenoprotein M
Gene Aliases
I79_017430
Chromosome Location
Chr.Un|NW_003614682: 385238 - 388082 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256844.1 NP_001243773.1 3 - 4 425 69
GenBank mRNA JI875827.1 - 3 - 4 266 69
JP039203.1 - 3 - 4 266 69

View Full Product Details