Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
100689412
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
LDLF
Chromosome Location
Chr.Un|NW_003613716: 627263 - 638204 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246677.1 NP_001233606.1 3 - 4 330 66
GenBank mRNA JI891514.1 - 3 - 4 337 66
JP045922.1 - 3 - 4 337 66
Z32554.1 - 3 - 4 330 66

View Full Product Details