Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myl2
Entrez Gene ID
100773969
Gene Name
myosin, light chain 2, regulatory, cardiac, slow
Gene Aliases
I79_004267
Chromosome Location
Chr.Un|NW_003613692: 1199988 - 1205455 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007627770.1 XP_007625960.1 4 - 5 341 84

View Full Product Details