Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psg9
Entrez Gene ID
100771218
Gene Name
pregnancy specific beta-1-glycoprotein 9
Gene Aliases
I79_016285
Chromosome Location
Chr.Un|NW_003614529: 568866 - 571666 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649005.1 XP_007647195.1 1 - 2 106 93

View Full Product Details