Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam19a4
Entrez Gene ID
100760241
Gene Name
family with sequence similarity 19 (chemokine (C-C motif)-like), member A4
Gene Aliases
I79_000380
Chromosome Location
Chr.Un|NW_003613585: 5143974 - 5279934 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495223.2 XP_003495271.2 1 - 2 120 88

View Full Product Details